Ethyl-3-bromoimidazo[1,2-a]pyridine-2-carboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 3-bromoimidazo[1,2-a]pyridine-2-carboxylate

AS21159G1 AS21159G5
ACTIAS21159G1EA 68 GBP
ACTIAS21159G1 ACTIAS21159G5
Ethyl-3-bromoimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 3-bromoimidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₀H₉BrN₂O₂
MW: 269.1000 g/mol
MDL Number: MFCD05864806
CAS Number: 143982-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR