2-(4-Fluorophenyl)isonicotinic acid
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBYA-6142-1GEA
0
GBP
COBBYA-6142-1G
2-(4-Fluorophenyl)isonicotinic acid
2-(4-Fluorophenyl)isonicotinic acid
Formula:
C₁₂H₈FNO₂ |
MDL Number:
MFCD14666481 CAS Number: 883528-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...