2,8-Dimethylquinolin-4(1H)-one

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,8-Dimethylquinolin-4-ol, 2,8-Dimethyl-4-hydroxyquinoline

HC-6693-5G HC-6693-25G HC-6693-1G
COBBHC-6693-5GEA 0 GBP
COBBHC-6693-5G COBBHC-6693-25G COBBHC-6693-1G
2,8-Dimethylquinolin-4(1H)-one
2,8-Dimethylquinolin-4(1H)-one
Formula: C₁₁H₁₁NO
MW: 173.2111 g/mol
MDL Number: MFCD00100264
CAS Number: 15644-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR