3-(3-Fluorophenylcarbamoyl)phenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-(3-Fluorophenylcarbamoyl)benzeneboronic acid
BB-3176-5G
BB-3176-1G
COBBBB-3176-5GEA
0
GBP
COBBBB-3176-5G
COBBBB-3176-1G
3-(3-Fluorophenylcarbamoyl)phenylboronic acid
3-(3-Fluorophenylcarbamoyl)phenylboronic acid
Formula:
C₁₃H₁₁BFNO₃ MW: 259.04 g/mol |
MDL Number:
MFCD09027207 CAS Number: 874288-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...