5-Bromo-2-hydroxyisonicotinic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4140-1G CA-4140-5G
COBBCA-4140-1GEA 0 GBP
COBBCA-4140-1G COBBCA-4140-5G
5-Bromo-2-hydroxyisonicotinic acid
5-Bromo-2-hydroxyisonicotinic acid
Formula: C₆H₄BrNO₃
MW: 218.0049 g/mol
MDL Number: MFCD08689503
CAS Number: 913836-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR