Cyclohexylboronic acid pinacol ester
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Cyclohexylboronic acid Pinacol ester, 2-Cyclohexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, (4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohexane
PN-2645-5G
PN-2645-25G
PN-2645-1G
COBBPN-2645-5GEA
0
GBP
COBBPN-2645-5G
COBBPN-2645-25G
COBBPN-2645-1G
Cyclohexylboronic acid pinacol ester
Cyclohexylboronic acid pinacol ester
Formula:
C₁₂H₂₃BO₂ MW: 210.12 g/mol |
MDL Number:
MFCD04038749 CAS Number: 87100-15-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...