2-Bromo-N-cyclopropylbenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4800-5G CA-4800-25G CA-4800-1G
COBBCA-4800-5GEA 0 GBP
COBBCA-4800-5G COBBCA-4800-25G COBBCA-4800-1G
2-Bromo-N-cyclopropylbenzamide
2-Bromo-N-cyclopropylbenzamide
Formula: C₁₀H₁₀BrNO
MDL Number: MFCD00740168
CAS Number: 88229-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR