1-Bromo-3-chloro-5-nitrobenzene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21740G1 AS21740G5
ACTIAS21740G1EA 0 GBP
ACTIAS21740G1 ACTIAS21740G5
1-Bromo-3-chloro-5-nitrobenzene
1-Bromo-3-chloro-5-nitrobenzene
Formula: C₆H₃BrClNO₂
MW: 236.4500 g/mol
MDL Number: MFCD08275708
CAS Number: 219817-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR