4-(Carboxymethyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(4-Boronophenyl)acetic acid

BB-3223-1G BB-3223-250M
COBBBB-3223-1GEA 0 GBP
COBBBB-3223-1G COBBBB-3223-250M
4-(Carboxymethyl)phenylboronic acid
4-(Carboxymethyl)phenylboronic acid
Formula: C₈H₉BO₄
MW: 179.9700 g/mol
MDL Number: MFCD03425919
CAS Number: 90111-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR