Tissue grinders, Mini series
Supplier: GPE Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GLPR20404EA 73.4 GBP
GLPR20404 GLPR20403 GLPR20402 GLPR20406 GLPR20405
Tissue grinders, Mini series
Homogenisers Tissue Grinders
These mini homogenisers are made of high quality borosilicate glass and are ideal for micro tissue work. The mortar is made from clear precision glass and pestles have ground glass working length. Mini homogenisers are available with or without handles.
Grinding clearance pestle and mortar: 0,152 to 0,254 mm.