Alizarin red S (sodium salt) solution Reag. Ph. Eur. 1002601

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Alizarin Red S sodiumSodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulphonateC.I. 58005Alizarin sulphonic acid disodium salt

87763.180EA 106 GBP
87763.180
Alizarin red S (sodium salt) solution Reag. Ph. Eur. 1002601
Alizarin Red S (sodium salt)

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: C₁₄H₇NaO₇S
MW: 342.26 g/mol
MDL Number: MFCD00013049
CAS Number: 130-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR