4-Mercaptobenzoic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Thiobenzoic acid

M-Clarity™ quality level = MQ100

662534-25G 662534-5G
SIAL662534-25GEA 279 GBP
SIAL662534-25G SIAL662534-5G
4-Mercaptobenzoic acid, Sigma-Aldrich®
4-Mercaptobenzoic acid
Formula: C₇H₆O₂S
MW: 154.1863 g/mol
MDL Number: MFCD00016617
CAS Number: 1074-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR