2-(3,4-Dimethoxyphenyl)ethyl isothiocyanate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D4650.10 D4650.50
XXXXD4650.10EA 0 GBP
XXXXD4650.10 XXXXD4650.50
2-(3,4-Dimethoxyphenyl)ethyl isothiocyanate
2-(3,4-Dimethoxyphenyl)ethyl isothiocyanate
Formula: C₁₁H₁₃NO₂S
MW: 223.30 g/mol
MDL Number: MFCD00041366
CAS Number: 21714-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR