2-(4-Chlorophenyl)ethyl isothiocyanate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

C2687.1 C2687.5
XXXXC2687.1EA 0 GBP
XXXXC2687.1 XXXXC2687.5
2-(4-Chlorophenyl)ethyl isothiocyanate
2-(4-Chlorophenyl)ethyl isothiocyanate
Formula: C₉H₈ClNS
MW: 197.69 g/mol
MDL Number: MFCD00041127
CAS Number: 17608-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR