Methyl-2,3,4-trifluorobenzoate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 2,3,4-trifluorobenzoate

021973-5G 021973-25G
FLUO021973-5GEA 0 GBP
FLUO021973-5G FLUO021973-25G
Methyl-2,3,4-trifluorobenzoate
Methyl 2,3,4-trifluorobenzoate
Formula: C₈H₅F₃O₂
MDL Number: MFCD06201866
CAS Number: 773873-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR