Methyl-4-isothiocyanatobenzoate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(Methoxycarbonyl)phenyl isothiocyanate

M1368-E.5 M1368-E.25
XXXXM1368-E.5EA 0 GBP
XXXXM1368-E.5 XXXXM1368-E.25
Methyl-4-isothiocyanatobenzoate
Methyl 4-isothiocyanatobenzoate
Formula: C₉H₇NO₂S
MW: 193.23 g/mol
MDL Number: MFCD00041099
CAS Number: 3662-78-0
EINECS: 240-164-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR