Ethyl-4-isothiocyanatobenzoate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(Ethoxycarbonyl)phenyl isothiocyanate

E1474-G.5 E1474-G.25
XXXXE1474-G.5EA 0 GBP
XXXXE1474-G.5 XXXXE1474-G.25
Ethyl-4-isothiocyanatobenzoate
Ethyl 4-isothiocyanatobenzoate
Formula: C₁₀H₉NO₂S
MW: 207.25 g/mol
MDL Number: MFCD00022058
CAS Number: 1205-06-7
EINECS: 214-880-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR