Methyl-3-isothiocyanatobenzoate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(Methoxycarbonyl)phenyl isothiocyanate

M1368-D.5 M1368-D.25
XXXXM1368-D.5EA 0 GBP
XXXXM1368-D.5 XXXXM1368-D.25
Methyl-3-isothiocyanatobenzoate
Methyl 3-isothiocyanatobenzoate
Formula: C₉H₇NO₂S
MDL Number: MFCD00041080
CAS Number: 3125-66-4
EINECS: 240-164-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR