Methyl-3-isothiocyanatobenzoate ≥99%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-(Methoxycarbonyl)phenyl isothiocyanate
L10476.06EA
0
GBP
L10476.06
Methyl-3-isothiocyanatobenzoate ≥99%
Methyl 3-isothiocyanatobenzoate
Formula:
C₉H₇NO₂S |
MDL Number:
MFCD00041080 CAS Number: 3125-66-4 EINECS: 240-164-2 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...