Ethyl-3-isothiocyanatobenzoate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

E1474-F.5 E1474-F.25
XXXXE1474-F.5EA 0 GBP
XXXXE1474-F.5 XXXXE1474-F.25
Ethyl-3-isothiocyanatobenzoate
Ethyl 3-isothiocyanatobenzoate
Formula: C₁₀H₉NO₂S
MDL Number: MFCD00060375
CAS Number: 3137-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR