Methyl-2,3-dimethylbenzoate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl 2,3-dimethylbenzoate
AS21511G25
AS21511G5
ACTIAS21511G25EA
191
GBP
ACTIAS21511G25
ACTIAS21511G5
Methyl-2,3-dimethylbenzoate
Methyl 2,3-dimethylbenzoate
Formula:
C₁₀H₁₂O₂ MW: 164.2000 g/mol |
MDL Number:
MFCD06203773 CAS Number: 15012-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...