Ethyl-2,3-dimethylbenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21512G25 AS21512G5
ACTIAS21512G25EA 218 GBP
ACTIAS21512G25 ACTIAS21512G5
Ethyl-2,3-dimethylbenzoate
Ethyl 2,3-dimethylbenzoate
Formula: C₁₁H₁₄O₂
MDL Number: MFCD00017523
CAS Number: 104175-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR