4-(Diethylamino)phenyl isothiocyanate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D2743.5 D2743.25
XXXXD2743.5EA 0 GBP
XXXXD2743.5 XXXXD2743.25
4-(Diethylamino)phenyl isothiocyanate
4-(Diethylamino)phenyl isothiocyanate
Formula: C₁₁H₁₄N₂S
MW: 206.31 g/mol
MDL Number: MFCD00041094
CAS Number: 84381-54-4
EINECS: 218-360-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR