3-(4-Morpholino)propyl isothiocyanate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

M4075.10 M4075.50
XXXXM4075.10EA 0 GBP
XXXXM4075.10 XXXXM4075.50
3-(4-Morpholino)propyl isothiocyanate
3-(4-Morpholino)propyl isothiocyanate
Formula: C₈H₁₄N₂OS
MW: 186.28 g/mol
MDL Number: MFCD00041222
CAS Number: 32813-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR