2-(4-Morpholino)ethyl isothiocyanate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

M4065.10 M4065.50
XXXXM4065.10EA 0 GBP
XXXXM4065.10 XXXXM4065.50
2-(4-Morpholino)ethyl isothiocyanate
2-(4-Morpholino)ethyl isothiocyanate
Formula: C₇H₁₂N₂OS
MW: 172.25 g/mol
MDL Number: MFCD00041221
CAS Number: 63224-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR