2-(4-Morpholino)ethyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009544-1G
009544-5G
009544-25G
FLUO009544-1GEA
0
GBP
FLUO009544-1G
FLUO009544-5G
FLUO009544-25G
2-(4-Morpholino)ethyl isothiocyanate
2-(4-Morpholino)ethyl isothiocyanate
Formula:
C₇H₁₂N₂OS MW: 172.25 g/mol |
MDL Number:
MFCD00041221 CAS Number: 63224-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...