2-(1-Piperidino)ethyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009568-1G 009568-5G
FLUO009568-1GEA 0 GBP
FLUO009568-1G FLUO009568-5G
2-(1-Piperidino)ethyl isothiocyanate
2-(1-Piperidino)ethyl isothiocyanate
Formula: C₈H₁₄N₂S
MW: 170.28 g/mol
MDL Number: MFCD00041232
CAS Number: 32813-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR