2-Pyrid-3-ylbenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(Pyridin-3-yl)benzoic acid

011853-500MG 011853-1G
FLUO011853-500MGEA 0 GBP
FLUO011853-500MG FLUO011853-1G
2-Pyrid-3-ylbenzoic acid
2-Pyrid-3-ylbenzoic acid
Formula: C₁₂H₉NO₂
MW: 199.2054 g/mol
MDL Number: MFCD03426511
CAS Number: 134363-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR