Water with 0,1% (v/v) trifluoroacetic acid for LC-MS, Pierce™

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PIER85172EA 115 GBP
PIER85172 PIER85173
Water with 0,1% (v/v) trifluoroacetic acid for LC-MS, Pierce™
Water with 0,1% (v/v) trifluoroacetic acid

Pierce™ 0.1% Trifluoroacetic Acid (v/v) in Water, LC-MS Grade is a preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.


  • Tested more than 20 ways to ensure maximal LC-MS sensitivity
  • Processed, filtered and sealed to extend LC-MS column life
  • Packaged to eliminate variability and reduce handling


Caution: For Research Use Only. Not for use in diagnostic procedures.

Storage Temperature: Ambient
EINECS: 200-929-3
https://uk.vwr.com/stibo/web/std.lang.all/83/92/728392.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR