2-Naphthyl boronic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Naphthaleneboronic acid, Naphthalene-2-boronic acid, 2-Naphthyl boric acid

AS21476G100 AS21476G25
ACTIAS21476G100EA 123 GBP
ACTIAS21476G100 ACTIAS21476G25
2-Naphthyl boronic acid
2-Naphthyl boronic acid
Formula: C₁₀H₉BO₂
MW: 171.99 g/mol
MDL Number: MFCD00236051
CAS Number: 32316-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR