4-Chloro-2-methoxyaniline hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Chloro-o-anisidine hydrochloride

AN-1484-5G AN-1484-1G
COBBAN-1484-5GEA 0 GBP
COBBAN-1484-5G COBBAN-1484-1G
4-Chloro-2-methoxyaniline hydrochloride
4-Chloro-2-methoxyaniline hydrochloride
Formula: C₇H₉Cl₂NO
MW: 194.06 g/mol
MDL Number: MFCD06801386
CAS Number: 861299-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR