4-Amino-1-naphthaleneboronic acid pinacol ester
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-1-amine
PN-6206-1G
PN-6206-250M
COBBPN-6206-1GEA
0
GBP
COBBPN-6206-1G
COBBPN-6206-250M
4-Amino-1-naphthaleneboronic acid pinacol ester
4-Amino-1-naphthaleneboronic acid pinacol ester
Formula:
C₁₆H₂₀BNO₂ MW: 269.1465 g/mol |
MDL Number:
MFCD15143602 CAS Number: 1218790-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...