2-Naphthyl boronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene, 4,4,5,5-Tetramethyl-2-(2-naphthyl)-1,3,2-dioxaborolane, Naphthalene-2-boronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-(naphthalen-2-yl)-1,3,2-dioxaborolane

PN-2672-5G PN-2672-25G PN-2672-1G
COBBPN-2672-5GEA 0 GBP
COBBPN-2672-5G COBBPN-2672-25G COBBPN-2672-1G
2-Naphthyl boronic acid pinacol ester
2-Naphthyl boronic acid pinacol ester
Formula: C₁₆H₁₉BO₂
MW: 254.14 g/mol
MDL Number: MFCD05663868
CAS Number: 256652-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR