1-Naphthyl boronic acid pinacol ester
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Naphthalene-1-boronic acid Pinacol ester, 1-Naphthalenboronic acid, pinacol ester, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene, 4,4,5,5-Tetramethyl-2-(1-naphthyl)-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-(naphthalen-1-yl)-1,3,2-dioxaborolane
PN-2671-25G
PN-2671-5G
COBBPN-2671-25GEA
0
GBP
COBBPN-2671-25G
COBBPN-2671-5G
1-Naphthyl boronic acid pinacol ester
1-Naphthyl boronic acid pinacol ester
Formula:
C₁₆H₁₉BO₂ MW: 254.1319 g/mol |
MDL Number:
MFCD05663869 CAS Number: 68716-52-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...