2-Fluoro-4-thiocyanatoaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1402-5G AN-1402-1G
COBBAN-1402-5GEA 0 GBP
COBBAN-1402-5G COBBAN-1402-1G
2-Fluoro-4-thiocyanatoaniline
2-Fluoro-4-thiocyanatoaniline
Formula: C₇H₅FN₂S
MDL Number: MFCD04617227
CAS Number: 14512-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR