3-Bromo-4-thiocyanatoaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1411-5G
AN-1411-1G
COBBAN-1411-5GEA
0
GBP
COBBAN-1411-5G
COBBAN-1411-1G
3-Bromo-4-thiocyanatoaniline
3-Bromo-4-thiocyanatoaniline
Formula:
C₇H₅BrN₂S |
MDL Number:
MFCD11855822 CAS Number: 16582-78-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...