2-Bromo-5-fluoro-4-thiocyanatoaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1398-5G AN-1398-1G
COBBAN-1398-5GEA 0 GBP
COBBAN-1398-5G COBBAN-1398-1G
2-Bromo-5-fluoro-4-thiocyanatoaniline
2-Bromo-5-fluoro-4-thiocyanatoaniline
Formula: C₇H₄BrFN₂S
MW: 247.0875 g/mol
MDL Number: MFCD11855815
CAS Number: 1133115-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR