4-Methyl-1-pentenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-Methylpent-1-en-1-yl)boronic acid

BB-8321-1G BB-8321-250M
COBBBB-8321-1GEA 0 GBP
COBBBB-8321-1G COBBBB-8321-250M
4-Methyl-1-pentenylboronic acid
4-Methyl-1-pentenylboronic acid
Formula: C₆H₁₃BO₂
MW: 127.98 g/mol
MDL Number: MFCD01074680
CAS Number: 214907-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR