1H-Pyrazole-3-boronic acid hydrate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

720429-250MG 720429-1G
SIAL720429-250MGEA 139 GBP
SIAL720429-250MG SIAL720429-1G
1H-Pyrazole-3-boronic acid hydrate, Sigma-Aldrich®
1H-Pyrazole-3-boronic acid hydrate
Formula: C₃H₅BN₂O₂·H₂O
MW: 111.90 g/mol
MDL Number: MFCD11044442
CAS Number: 376584-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR