5,6,7,8-Tetrahydro-1-naphthaleneboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (5,6,7,8-Tetrahydronaphthalen-1-yl)boronic acid

BB-9052-1G BB-9052-250M
COBBBB-9052-1GEA 0 GBP
COBBBB-9052-1G COBBBB-9052-250M
5,6,7,8-Tetrahydro-1-naphthaleneboronic acid
5,6,7,8-Tetrahydro-1-naphthaleneboronic acid
Formula: C₁₀H₁₃BO₂
MW: 176.200 g/mol
MDL Number: MFCD08701755
CAS Number: 371765-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR