5,6,7,8-Tetrahydro-2-naphthaleneboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PN-6250-1G PN-6250-250M
COBBPN-6250-1GEA 0 GBP
COBBPN-6250-1G COBBPN-6250-250M
5,6,7,8-Tetrahydro-2-naphthaleneboronic acid pinacol ester
5,6,7,8-Tetrahydro-2-naphthaleneboronic acid pinacol ester
Formula: C₁₆H₂₃BO₂
MDL Number: MFCD15143611
CAS Number: 627526-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR