6-Bromoimidazo[1,2-a]pyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI-1235-1G
HI-1235-5G
COBBHI-1235-1GEA
0
GBP
COBBHI-1235-1G
COBBHI-1235-5G
6-Bromoimidazo[1,2-a]pyridine
6-Bromoimidazo[1,2-a]pyridine
Formula:
C₇H₅BrN₂ MW: 197.03 g/mol |
MDL Number:
MFCD06496200 CAS Number: 6188-23-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...