3-Bromo-6-chloroimidazo[1,2-b]pyridazine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI-1277-5G
HI-1277-25G
HI-1277-1G
COBBHI-1277-5GEA
0
GBP
COBBHI-1277-5G
COBBHI-1277-25G
COBBHI-1277-1G
3-Bromo-6-chloroimidazo[1,2-b]pyridazine
3-Bromo-6-chloroimidazo[1,2-b]pyridazine
Formula:
C₆H₃BrClN₃ MW: 232.47 g/mol |
MDL Number:
MFCD09027274 CAS Number: 13526-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...