1,4-Dioxaspiro[4.5]decan-8-one
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20600G100
AS20600G25
ACTIAS20600G100EA
187
GBP
ACTIAS20600G100
ACTIAS20600G25
1,4-Dioxaspiro[4.5]decan-8-one
1,4-Dioxaspiro[4.5]decan-8-one
Formula:
C₈H₁₂O₃ |
MDL Number:
MFCD00010214 CAS Number: 4746-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...