Benzyl glycidyl ether, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-((Benzyloxy)methyl)oxirane

M-Clarity™ quality level = MQ100

469785-25ML 469785-5ML
SIAL469785-25MLEA 169 GBP
SIAL469785-25ML SIAL469785-5ML
Benzyl glycidyl ether, Sigma-Aldrich®
Benzyl glycidyl ether
Formula: C₁₀H₁₂O₂
MW: 164.2011 g/mol
MDL Number: MFCD00068664
CAS Number: 2930-05-4
EINECS: 220-899-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR