Furan-3-boronic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Furanboronic acid, 3-Furanylboronic acid
AS35199G1
AS35199G5
ACTIAS35199G1EA
18
GBP
ACTIAS35199G1
ACTIAS35199G5
Furan-3-boronic acid
Furan-3-boronic acid
Formula:
C₄H₅BO₃ MW: 111.89 g/mol |
MDL Number:
MFCD01319007 CAS Number: 55552-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...