1-Chlorobenzotriazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS51508G25 AS51508G5
ACTIAS51508G25EA 130 GBP
ACTIAS51508G25 ACTIAS51508G5
1-Chlorobenzotriazole
1-Chlorobenzotriazole
Formula: C₆H₄ClN₃
MDL Number: MFCD00054541
CAS Number: 21050-95-3
EINECS: 202-378-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR