1,1,1,3,3,3-Hexafluoroisopropyltoluene-4-sulphonate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,1,1,3,3,3-Hexafluoropropan-2-yl 4-methylbenzenesulphonate

001511-25G 001511-100G
FLUO001511-25GEA 0 GBP
FLUO001511-25G FLUO001511-100G
1,1,1,3,3,3-Hexafluoroisopropyltoluene-4-sulphonate
1,1,1,3,3,3-Hexafluoroisopropyl toluene-4-sulphonate
Formula: C₁₀H₈F₆O₃S
MW: 322.23 g/mol
MDL Number: MFCD00039262
CAS Number: 67674-48-0
EINECS: 266-887-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR