2-Fluoro-4-iodobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS12082G1 AS12082G5
ACTIAS12082G1EA 0 GBP
ACTIAS12082G1 ACTIAS12082G5
2-Fluoro-4-iodobenzaldehyde
2-Fluoro-4-iodobenzaldehyde
Formula: C₇H₄FIO
MW: 250.01 g/mol
MDL Number: MFCD07782056
CAS Number: 699016-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR