2-Amino-6-methoxy-3-nitropyridine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Methoxy-3-nitropyridin-2-amine

M-Clarity™ quality level = MQ100

704598-1G 704598-5G
SIAL704598-1GEA 35.8 GBP
SIAL704598-1G SIAL704598-5G
2-Amino-6-methoxy-3-nitropyridine, Sigma-Aldrich®
2-Amino-6-methoxy-3-nitropyridine
Formula: C₆H₇N₃O₃
MW: 169.14 g/mol
MDL Number: MFCD03206481
CAS Number: 73896-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR